Assay Details
| Gene Symbol: Szrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114599.2 | NP_001108071.1 | 4 | 1283 | 97 | |
| NR_102345.1 | 3 | 1233 | 97 | |||
| XM_006239264.2 | XP_006239326.1 | 4 | 1311 | 97 | ||
| GenBank mRNA | BC083784.1 | - | 5 | 1365 | 97 | |
| FQ222061.1 | - | 3 | 97 | |||
More Information
Set Membership: |
|

Szrd1
Genomic Map