Assay Details
Gene Symbol: Isca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181626.3 | NP_853657.1 | 4 | 427 | 87 | |
GenBank mRNA | AF508158.1 | - | 4 | 393 | 87 | |
BC070929.1 | - | 4 | 427 | 7096424 | 87 | |
BC078677.1 | - | 4 | 437 | 7133788 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
