Assay Details
| Gene Symbol: Sdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013026.2 | NP_037158.1 | 5 | 1560 | 65 | |
| GenBank mRNA | BC061834.1 | - | 5 | 1560 | 5600086 | 65 |
| M81785.1 | - | 5 | 1575 | 65 | ||
| S61865.1 | - | 5 | 1575 | 65 | ||
More Information
Set Membership: |
|

Sdc1
Genomic Map