Assay Details
| Gene Symbol: Nkx2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006240118.2 | XP_006240180.1 | 1 | 50 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006240118.2 transcripts.
Set Membership: |
|

Nkx2-1
Genomic Map