Assay Details
| Gene Symbol: Chst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241200.2 | XP_006241262.1 | 1 | 530 | 81 | |
| XM_008765238.1 | XP_008763460.1 | 2 | 552 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006241200.2, XM_008765238.1 transcripts.
Set Membership: |
|

Chst11
Genomic Map