Assay Details
| Gene Symbol: LOC499182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100474.1 | - | 3 | 4358 | 91 | |
| Gene Symbol: Crp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY310153.1 | - | 746 | 91 | ||
| AY318961.1 | - | 746 | 91 | |||
| AY321318.1 | - | 743 | 91 | |||
| AY321324.1 | - | 746 | 91 | |||
| AY325196.1 | - | 746 | 91 | |||
| AY325218.1 | - | 746 | 91 | |||
| AY325232.1 | - | 746 | 91 | |||
| Gene Symbol: LOC100362110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S80119.1 | - | 720 | 91 | ||
| Gene Symbol: LOC499355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100475.1 | - | 3 | 4355 | 91 | |
| Gene Symbol: LOC501503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100477.1 | - | 2 | 4359 | 91 | |
| Gene Symbol: Lrrgt00014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U83119.1 | - | 1 | 2662 | 91 | |
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY539942.1 | - | 1957 | 91 | ||
More Information
Set Membership: |
|

LOC499182
Genomic Map