Assay Details
| Gene Symbol: Bcl2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033670.1 | NP_001028842.1 | 3 | 2238 | 90 | |
| NM_001033671.1 | NP_001028843.1 | 2 | 1983 | 90 | ||
| XM_006235265.2 | XP_006235327.1 | 3 | 3050 | 90 | ||
| GenBank mRNA | BC094213.1 | - | 3 | 2238 | 7314572 | 90 |
More Information
Set Membership: |
|

Bcl2l1
Genomic Map