Assay Details
| Gene Symbol: LOC100911148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229637.2 | XP_006229699.1 | 4 | 2481 | 79 | |
| Gene Symbol: LOC690082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759620.1 | XP_008757842.1 | 2 | 3357 | 79 | |
More Information
Set Membership: |
|

LOC100911148
Genomic Map