Assay Details
| Gene Symbol: LOC100910502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771840.1 | XP_008770062.1 | 2 | 522 | 138 | |
| Gene Symbol: LOC100912428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770132.1 | XP_008768354.1 | 2 | 552 | 138 | |
| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271366.1 | NP_001258295.1 | 13 | 8640 | 138 | |
| XM_006230453.2 | XP_006230515.1 | 12 | 8989 | 138 | ||
More Information
Set Membership: |
|

LOC100910502
Genomic Map