Assay Details
| Gene Symbol: Dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001053826.4 | XP_001053826.1 | 1 | 172 | 91 | |
| XM_006242286.2 | XP_006242348.1 | 1 | 172 | 91 | ||
More Information
Set Membership: |
|

Dbx2
Genomic Map