Assay Details
| Gene Symbol: LOC102551453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765863.1 | XP_008764085.1 | 1 | 338 | 92 | |
| Gene Symbol: Krt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183333.1 | NP_899162.1 | 450 | 92 | ||
| GenBank mRNA | AY342389.1 | - | 450 | 92 | ||
| BC062086.1 | - | 313 | 5599969 | 92 | ||
More Information
Set Membership: |
|

LOC102551453
Genomic Map