Assay Details
| Gene Symbol: Aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040156.2 | NP_001035246.2 | 2 | 481 | 128 | |
| XM_006243672.2 | XP_006243734.1 | 1 | 236 | 128 | ||
| XM_008766543.1 | XP_008764765.1 | 1 | 665 | 128 | ||
| XM_008766544.1 | XP_008764766.1 | 1 | 665 | 128 | ||
More Information
Set Membership: |
|

Aste1
Genomic Map