Assay Details
Gene Symbol: LOC103689995 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765895.1 | XP_008764117.1 | 595 | 81 | ||
XM_008765896.1 | XP_008764118.1 | 595 | 81 |
Gene Symbol: Dync2h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023024.1 | NP_075413.1 | 43 - 44 | 7532 | 81 | |
GenBank mRNA | AB041881.1 | - | 43 - 44 | 7532 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
