Assay Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025123.3 | NP_001020294.3 | 8 | 3067 | 53 | |
| GenBank mRNA | BC100617.1 | - | 8 | 3066 | 7934552 | 53 |
| FQ211817.1 | - | 7 | 2771 | 53 | ||
More Information
Set Membership: |
|

Kdelc2
Genomic Map