Assay Details
| Gene Symbol: Mrap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243487.2 | XP_006243549.1 | 6 | 2271 | 117 | |
| XM_006243488.2 | XP_006243550.1 | 5 | 2126 | 117 | ||
| XM_008766452.1 | XP_008764674.1 | 6 | 2259 | 117 | ||
| XM_008766453.1 | XP_008764675.1 | 5 | 2159 | 117 | ||
More Information
Set Membership: |
|

Mrap2
Genomic Map