Assay Details
| Gene Symbol: Ccr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053960.3 | NP_446412.2 | 2 | 1012 | 89 | |
| GenBank mRNA | BC078756.1 | - | 1 | 947 | 7109679 | 89 |
| Y12009.1 | - | 2 | 1022 | 89 | ||
More Information
Set Membership: |
|

Ccr5
Genomic Map