Assay Details
| Gene Symbol: LOC103690063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008769965.1 | XP_008768187.1 | 1 - 2 | 626 | 118 | |
| Gene Symbol: Abcg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250626.2 | XP_006250688.1 | 1 - 2 | 104 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006250626.2, XM_008769965.1 transcripts.
Set Membership: |
|

LOC103690063
Genomic Map