Assay Details
| Gene Symbol: Ms4a13-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231105.2 | XP_006231167.1 | 1 - 2 | 247 | 84 | |
| XM_008760270.1 | XP_008758492.1 | 1 - 2 | 247 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006231105.2, XM_008760270.1 transcripts.
Set Membership: |
|

Ms4a13-ps1
Genomic Map