Assay Details
| Gene Symbol: LOC102551397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773039.1 | XP_008771261.1 | 391 | 107 | ||
| Gene Symbol: LOC102551192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773038.1 | XP_008771260.1 | 4 - 5 | 391 | 107 | |
More Information
Set Membership: |
|

LOC102551397
Genomic Map