Assay Details
Gene Symbol: Samt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001060573.5 | XP_001060573.3 | 2 - 3 | 265 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001060573.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
