Assay Details
| Gene Symbol: LOC681382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001061496.2 | XP_001061496.1 | 1 | 117 | 95 | |
| Gene Symbol: Cenpw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246319.1 | NP_001233248.1 | 1 | 95 | ||
| GenBank mRNA | BC129114.1 | - | 1 | 7112235 | 95 | |
More Information
Set Membership: |
|

LOC681382
Genomic Map