Assay Details
| Gene Symbol: Fam178a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134612.1 | NP_001128084.1 | 19 | 5635 | 123 | |
| XM_006231550.2 | XP_006231612.1 | 20 | 5791 | 123 | ||
| XM_006231551.2 | XP_006231613.1 | 20 | 5787 | 123 | ||
More Information
Set Membership: |
|

Fam178a
Genomic Map