Assay Details
Gene Symbol: Ott | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257440.2 | XP_006257502.1 | 2887 | 83 | ||
XM_008773481.1 | XP_008771703.1 | 2797 | 83 | |||
XM_008773482.1 | XP_008771704.1 | 2791 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
