Assay Details
| Gene Symbol: LOC100910506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006256650.1 | XP_006256712.1 | 13 | 2768 | 72 | |
| XM_006256651.1 | XP_006256713.1 | 12 | 2699 | 72 | ||
More Information
Set Membership: |
|

LOC100910506
Genomic Map