Assay Details
| Gene Symbol: LOC102549010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773213.1 | XP_008771435.1 | 7 - 8 | 1086 | 78 | |
| XM_008773214.1 | XP_008771436.1 | 6 - 7 | 939 | 78 | ||
More Information
Set Membership: |
|

LOC102549010
Genomic Map