Assay Details
| Gene Symbol: Ngfrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006257257.2 | XP_006257319.1 | 3 | 782 | 87 | |
| XM_006257258.2 | XP_006257320.1 | 3 | 696 | 87 | ||
| GenBank mRNA | AY833556.1 | - | 3 | 687 | 87 | |
| BC058503.1 | - | 3 | 786 | 6887969 | 87 | |
| BC166418.1 | - | 3 | 695 | 7314222 | 87 | |
More Information
Set Membership: |
|

Ngfrap1
Genomic Map