Assay Details
| Gene Symbol: Casp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760547.1 | XP_008758769.1 | 1 | 217 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008760547.1 transcripts.
Set Membership: |
|

Casp7
Genomic Map