Assay Details
| Gene Symbol: Magea10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229518.2 | XP_006229580.1 | 2 - 3 | 732 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229518.2 transcripts.
Set Membership: |
|

Magea10
Genomic Map