Assay Details
| Gene Symbol: LOC103694544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773682.1 | XP_008771904.1 | 7 - 8 | 840 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103694544| Gene Symbol: LOC103694544 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773682.1 | XP_008771904.1 | 7 - 8 | 840 | 68 | |
Set Membership: |
|