Assay Details
| Gene Symbol: Sgk494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246951.2 | XP_006247013.2 | 4 - 5 | 567 | 62 | |
| XM_008768145.1 | XP_008766367.1 | 4 - 5 | 567 | 62 | ||
| XM_008768146.1 | XP_008766368.1 | 3 - 4 | 483 | 62 | ||
| XM_008768147.1 | XP_008766369.1 | 4 - 5 | 567 | 62 | ||
More Information
Set Membership: |
|

Sgk494
Genomic Map