Assay Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768412.1 | XP_008766634.1 | 6 - 7 | 3093 | 61 | |
| GenBank mRNA | BC091694.1 | - | 5 - 6 | 1067 | 61 | |
More Information
Set Membership: |
|

Trim80
Genomic Map