Assay Details
| Gene Symbol: Map3k10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191621.3 | NP_001178550.1 | 1 | 203 | 124 | |
| XM_006228576.2 | XP_006228638.1 | 1 | 412 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191621.3, XM_006228576.2 transcripts.
Set Membership: |
|

Map3k10
Genomic Map