Assay Details
| Gene Symbol: LOC103693504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768542.1 | XP_008766764.1 | 1 | 3976 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768542.1 transcripts.
Set Membership: |
|

LOC103693504
Genomic Map