Assay Details
Gene Symbol: LOC100911399 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751031.3 | XP_003751079.2 | 1 - 2 | 596 | 50 |
Gene Symbol: Brwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751022.3 | XP_003751070.1 | 1 - 2 | 138 | 50 | |
XM_006248185.2 | XP_006248247.1 | 1 - 2 | 138 | 50 | ||
XM_006248186.2 | XP_006248248.1 | 1 - 2 | 141 | 50 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
