Assay Details
| Gene Symbol: LOC100911790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248417.1 | XP_006248479.1 | 3 - 4 | 210 | 93 | |
| Gene Symbol: Stfa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248416.2 | XP_006248478.1 | 2 - 3 | 241 | 93 | |
More Information
Set Membership: |
|

LOC100911790
Genomic Map