Assay Details
| Gene Symbol: Gmnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248531.2 | XP_006248593.1 | 1 - 2 | 203 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248531.2 transcripts.
Set Membership: |
|

Gmnc
Genomic Map