Assay Details
| Gene Symbol: RGD1564571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_221808.7 | XP_221808.5 | 1 - 2 | 130 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_221808.7 transcripts.
Set Membership: |
|

RGD1564571
Genomic Map