Assay Details
| Gene Symbol: RGD1564571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_221808.7 | XP_221808.5 | 5 - 6 | 567 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564571| Gene Symbol: RGD1564571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_221808.7 | XP_221808.5 | 5 - 6 | 567 | 93 | |
Set Membership: |
|