Assay Details
| Gene Symbol: Ywhag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249184.2 | XP_006249246.1 | 2 | 1894 | 92 | |
| GenBank mRNA | D17447.1 | - | 2 | 1729 | 92 | |
| S55305.1 | - | 2 | 1729 | 92 | ||
More Information
Set Membership: |
|

Ywhag
Genomic Map