Assay Details
| Gene Symbol: Ssh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249511.1 | XP_006249573.1 | 10 | 3881 | 120 | |
| XM_006249512.2 | XP_006249574.1 | 14 | 4273 | 120 | ||
More Information
Set Membership: |
|

Ssh1
Genomic Map