Assay Details
| Gene Symbol: Naa30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251813.2 | XP_006251875.1 | 1 | 195 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006251813.2 transcripts.
Set Membership: |
|

Naa30
Genomic Map