Assay Details
| Gene Symbol: LOC102550885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252233.2 | XP_006252295.1 | 1 - 2 | 81 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006252233.2 transcripts.
Set Membership: |
|

LOC102550885
Genomic Map