Assay Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108876.1 | NP_001102346.1 | 4 | 880 | 123 | |
| GenBank mRNA | FQ216033.1 | - | 4 | 650 | 123 | |
| FQ220497.1 | - | 4 | 748 | 123 | ||
More Information
Set Membership: |
|

Lsm1
Genomic Map