Assay Details
| Gene Symbol: LOC100361579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771615.1 | XP_008769837.1 | 1 - 2 | 154 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008771615.1 transcripts.
Set Membership: |
|

LOC100361579
Genomic Map