Assay Details
| Gene Symbol: Ovol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230794.2 | XP_006230856.1 | 1 | 746 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230794.2 transcripts.
Set Membership: |
|

Ovol1
Genomic Map