Assay Details
| Gene Symbol: Fam64a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113781.2 | NP_001107253.2 | 2 | 277 | 59 | |
| XM_006246759.2 | XP_006246821.1 | 2 | 387 | 59 | ||
| XM_006246760.2 | XP_006246822.1 | 2 | 363 | 59 | ||
| GenBank mRNA | BC158692.1 | - | 2 | 277 | 9022735 | 59 |
More Information
Set Membership: |
|

Fam64a
Genomic Map