Assay Details
| Gene Symbol: LOC100909950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254325.2 | XP_006254387.1 | 1 - 2 | 356 | 102 | |
| Gene Symbol: Pip4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053926.2 | NP_446378.1 | 2 - 3 | 341 | 102 | |
| GenBank mRNA | AB032899.1 | - | 2 - 3 | 341 | 102 | |
More Information
Set Membership: |
|

LOC100909950
Genomic Map