Assay Details
| Gene Symbol: Zscan30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254475.2 | XP_006254537.1 | 5 | 3264 | 93 | |
| XM_006254478.2 | XP_006254540.1 | 4 | 1770 | 93 | ||
| XM_006254479.2 | XP_006254541.1 | 5 | 3159 | 93 | ||
| XM_006254480.2 | XP_006254542.1 | 4 | 1678 | 93 | ||
| XM_006254481.2 | XP_006254543.1 | 5 | 1798 | 93 | ||
| XM_006254482.2 | XP_006254544.1 | 4 | 3011 | 93 | ||
| XM_008771999.1 | XP_008770221.1 | 4 | 6420 | 93 | ||
| XM_008772000.1 | XP_008770222.1 | 4 | 5059 | 93 | ||
| XM_008772001.1 | XP_008770223.1 | 4 | 3116 | 93 | ||
| XM_008772002.1 | XP_008770224.1 | 3 | 1322 | 93 | ||
More Information
Set Membership: |
|

Zscan30
Genomic Map