Assay Details
| Gene Symbol: Nefm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017029.2 | NP_058725.1 | 3 | 2939 | 136 | |
| XM_008770736.1 | XP_008768958.1 | 4 | 3276 | 136 | ||
| GenBank mRNA | Z12152.1 | - | 3 | 2942 | 136 | |
More Information
Set Membership: |
|

Nefm
Genomic Map