Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109625.2 | NP_001103095.2 | 10 | 2643 | 95 | |
| GenBank mRNA | BC166464.1 | - | 10 | 2358 | 9030793 | 95 |
| BG668882.1 | - | 1 | 73 | 95 | ||
| CA505328.1 | - | 1 | 95 | |||
| EX488132.1 | - | 1 | 95 | |||
More Information
Set Membership: |
|

Capza1
Genomic Map